missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
POLR2C Monoclonal antibody specifically detects POLR2C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
Specifications
| Antigen | POLR2C |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | SR1551 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200 |
| Formulation | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Gene Alias | DNA-directed RNA polymerase II 33 kDa polypeptide, DNA-directed RNA polymerase II subunit C, DNA-directed RNA polymerase II subunit RPB3, hRPB33, hsRPB3, polymerase (RNA) II (DNA directed) polypeptide C (33kD), polymerase (RNA) II (DNA directed) polypeptide C, 33kDa, RNA polymerase II subunit 3, RNA polymerase II subunit B3, RPB3, RPB31, RPB33 |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human POLR2C (Uniprot #: P19387) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?