missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530682
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431. Predicted reactivity: Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Gap-filling polymerase involved in repair of DNA double-strand breaks by non-homologous end joining (NHEJ). Participates in immunoglobulin (Ig) light chain gene rearrangement in V(D)J recombination.Specifications
POLM | |
Polyclonal | |
Unconjugated | |
POLM | |
AI462166; B230309I03Rik; DNA polymerase mu; DNA-directed DNA polymerase mu; DNA-directed DNA/RNA polymerase mu; Pol iota; pol Mu; Polm; polmu; polymerase (DNA directed), mu; polymerase (DNA) mu; polymerase (DNA-directed), mu; Tdt-N; Terminal transferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27434 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9NP87 | |
POLM | |
Recombinant fragment corresponding to a region within amino acids 49 and 268 of Human DNA polymerase mu | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |