missing translation for 'onlineSavingsMsg'
Learn More

Poliumoside, MedChemExpress

Product Code. p-200101197
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488828 10 mg 10mg
16424737 50 mg 50mg
16408966 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488828 Supplier Medchem Express Supplier No. HYN0033

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Poliumoside, a caffeoylated phenylpropanoid glycoside, is isolated from Brandisia hancei stems and leaves. Poliumoside is an advanced glycation end product (AGE) formation and rat lens aldose reductase (RLAR) inhibitor, with IC50s of 19.69 and 8.47 μM, respectively. Poliumoside also has antiinflammatory and antioxidant activity.

Poliumoside, a caffeoylated phenylpropanoid glycoside, is isolated from Brandisia hancei stems and leaves. Poliumoside is an advanced glycation end product (AGE) formation and rat lens aldose reductase (RLAR) inhibitor, with IC50s of 19.69 and 8.47 μM, respectively. Poliumoside also has antiinflammatory and antioxidant activity.

Specifications

Chemical Name or Material Poliumoside
CAS 94079-81-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C35H46O19
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (129.75 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (64.87 mM; Need ultrasonic)
SMILES OC1=C(O)C=CC(CCOC2C(C(OC3OC(C)C(O)C(O)C3O)C(OC(/C=C/C4=CC=C(O)C(O)=C4)=O)C(COC5OC(C)C(O)C(O)C5O)O2)O)=C1
Molecular Weight (g/mol) 770.73
Formula Weight 770.73
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.