Learn More
Ekastu Safety™ Polimask 330/2 Half Mask
Product Code. 10067850
Click to view available options
Product Type:
Half Mask
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 10067850
Brand: Ekastu Safety™ 466605
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Includes standard thread connection according to DIN EN 148-1. EKASTU Safety™ Polimask 330/2 Half Mask is made of soft EPDM and specially designed for spectacle wearers.
  • With standard thread connection according to DIN EN 148-1
  • Mask body with exhalation valve and support under the chin
  • Made of soft EPDM
  • Owing to the special design the mask is suitable for spectacle wearers
  • The connector allows the half mask to be used with screw filters up to 300g, pressure hose devices and turbo unit respiratory systems
Specifications
Certifications/Compliance DIN EN 148-1
Material Soft EPDM
Product Type Half Mask
Facepiece Type Half mask
For Use With (Equipment) Can be used with screw filters up to 300g, pressure hose devices and turbo unit respiratory systems
Model Polimask 330
Quantity 1 Ea.