Learn More
Thermo Scientific™ Point of Dispense Filters for Barnstead™ Point of Use Water Purification Systems
Sterile filter for use with Barnstead™ water purification systems; can be autoclaved up to 5 times.
Brand:  Thermo Scientific™ 50157375
266.76 EUR valid until 2024-02-28
Use promo code "21740" to get your promotional price.
Additional Details : Weight : 0.06820kg
 View more versions of this product
Product Code. 15950206
  • 280.80 € / 1 unit
Estimated Shipment: 21-03-2024
to see stock.
Add to basket
Description
Description
Reduce bacteria using the Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems.
Replacement consumable for the following Barnstead water purification systems:
  • GenPure™
  • GenPure™ Pro
  • GenPure™ xCAD
  • LabTower™ EDI
  • LabTower™ TII
  • MicroPure™
  • Smart2Pure™
  • Smart2Pure™ Pro
Specifications
Specifications
Point of Dispense filter
Sterile Filter, 0.1μm
Water Purification System
Smart2Pure Pro
Recommendations
Recommendations