missing translation for 'onlineSavingsMsg'
Learn More

Podoplanin Antibody (PDPN/8950R), Novus Biologicals™

Rabbit Monoclonal Antibody

268.00 € - 496.00 €

Specifications

Antigen Podoplanin
Clone PDPN/8950R
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18699314
View Documents
Novus Biologicals
NBP3-23709-20ug
20 μg
268.00 €
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18631784
View Documents
Novus Biologicals
NBP3-23709-100ug
100 μg
496.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Podoplanin Monoclonal antibody specifically detects Podoplanin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Podoplanin
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Core ESC Like Genes, Signal Transduction, Stem Cell Markers
10 mM PBS with 0.05% BSA
10630
IgG κ
Protein A or G purified
PDPN/8950R
Immunohistochemistry-Paraffin : 1-2 μg/mL
Monoclonal
Purified
RUO
Human
36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha
Recombinant fragment of human Podoplanin protein (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.