missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ POC5 Recombinant Protein Antigen
Highly purified and high bioactivity. Generating reliable and reproducible results.
Applications: Antibody Competition
237.57 €
Specifications
Gene ID (Entrez) | 134359 |
---|---|
Species | Human |
Purity | >80% by SDS-PAGE and Coomassie blue staining |
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
For Use With (Application) | Antibody Competition |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18203899
|
Novus Biologicals™
NBP2-76534PEP |
Amino Acid Sequence: KWEEYEELLHYAIVTPNIEPCASQSSHPKGELVPDVRISTIHDILHSQGNNSEVRETAIEVGKGCDFHISSHSKTDESSPVLSPRKPSHPVMD |
237.57 €
100µL |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18219428
|
Novus Biologicals™
NBP2-76555PEP |
Amino Acid Sequence: YVPRVVTSAQQKAGRTITARITGRCDFASKNRISSSLAIMGVSPPMSSVVVEKHHPVTVQTIPQATAAKYPRTIHPESSTSASRSLGTRSAHTQSLTSVH |
237.57 €
100µL |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human POC5The POC5 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
134359 | |
>80% by SDS-PAGE and Coomassie blue staining | |
Antibody Competition | |
Unconjugated | |
100 μL | |
E. coli | |
PBS and 1M Urea, pH 7.4. |
Human | |
Store at -20°C. Avoid freeze-thaw cycles. | |
POC5 | |
28 kDa | |
RUO | |
POC5 |