missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNPO Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15552-1-AP-20UL
This item is not returnable.
View return policy
Description
The enzyme encoded by this gene catalyzes the terminal, rate-limiting step in the synthesis of pyridoxal 5′-phosphate, also known as vitamin B6. Vitamin B6 is a required co-factor for enzymes involved in both homocysteine metabolism and synthesis of neurotransmitters such as catecholamine.Specifications
| PNPO | |
| Polyclonal | |
| Unconjugated | |
| PNPO | |
| PDXPO, PNPO, Pyridoxamine phosphate oxidase, Pyridoxine 5 phosphate oxidase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 103711, 55163, 64533 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
| 0.55 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88794, Q91XF0, Q9NVS9 | |
| PNPO | |
| PNPO Fusion Protein Ag7914 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction