missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNPLA6 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-17292-100UL
This item is not returnable.
View return policy
Description
PNPLA6 Polyclonal antibody specifically detects PNPLA6 in Human samples. It is validated for Immunofluorescence
Specifications
| PNPLA6 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
| EC 3.1.1.5, neuropathy target esterase, NTENTEMND, patatin-like phospholipase domain containing 6, Patatin-like phospholipase domain-containing protein 6, SPG39, sws | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: SYCEDESATGGCPFGPYQGRQTSSIFEAAKQELAKLMRIEDPSLLNSRVLLHHAKAGTIIARQGDQDVSLHFVLWG | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10908 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction