missing translation for 'onlineSavingsMsg'
Learn More

PNPLA5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16521402
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16521402 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16521402 Supplier Invitrogen Supplier No. PA598575

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PNPLA5 Polyclonal Antibody

PNPLA5 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen PNPLA5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PNPLA5
Gene Accession No. Q7Z6Z6
Gene Alias dJ388M5; dJ388M5.4; GS2L; GS2-like protein; patatin like phospholipase domain containing 5; patatin-like phospholipase domain containing 5; patatin-like phospholipase domain-containing protein 5; PNPLA5
Gene Symbols PNPLA5
Host Species Rabbit
Immunogen Recombinant Human Patatin-like phospholipase domain-containing protein 5 protein (90-205AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150379
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.