missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies PneumaCult™-Ex Medium

Product Code. 15376795
Encompass
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15376795 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15376795 Supplier STEMCELL Technologies Supplier No. 05008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Serum- and BPE-free medium for expansion of primary human airway epithelial cells

  • PneumaCult™-Ex is a defined, serum- and BPE-free cell culture medium that supports rapid expansion of human airway epithelial cells
  • Primary airway epithelial cells cultured in PneumaCult™-Ex expand rapidly over at least 3 passages while maintaining a cobblestone morphology and uniform expression of the basal cell markers p63 and p75NTR
  • Additionally, cells cultured in PneumaCult™-Ex can be differentiated to form a pseudostratified mucociliary epithelium when cultured at the air-liquid interface in PneumaCult™-ALI
  • Together, PneumaCult™-Ex and PneumaCult™-ALI constitute a fully integrated BPE-free culture system for in vitro human airway modeling
  • This robust and defined system is a valuable tool for basic respiratory research, toxicity studies, and drug development

Specifications

Cell Type Airway Cells
For Use With (Application) Cell Culture; Expansion; Maintenance
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.