missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PMPCB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598066
This item is not returnable.
View return policy
Description
PMPCB Polyclonal Antibody for Western Blot, IHC (P), ELISA
PMPCB is a member of the peptidase M16 family and encodes a protein with a zinc-binding motif. This protein is located in the mitochondrial matrix and catalyzes the cleavage of the leader peptides of precursor proteins newly imported into the mitochondria, though it only functions as part of a heterodimeric complex.
Specifications
| PMPCB | |
| Polyclonal | |
| Unconjugated | |
| PMPCB | |
| 3110004O18Rik; Beta-MPP; mitochondrial processing peptidase beta subunit; mitochondrial-processing peptidase subunit beta; MPP11; MPPB; MPPP52; P-52; peptidase (mitochondrial processing) beta; peptidase, mitochondrial processing beta subunit; PMPCB | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9512 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75439 | |
| PMPCB | |
| Recombinant Human Mitochondrial-processing peptidase subunit beta protein (110-380AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction