missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMCA1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-97754
This item is not returnable.
View return policy
Description
PMCA1 Polyclonal specifically detects PMCA1 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| PMCA1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| ATPase, Ca++ transporting, plasma membrane 1, EC 3.6.3, EC 3.6.3.8, plasma membrane calcium ATPase, Plasma membrane calcium ATPase isoform 1, plasma membrane calcium pump, Plasma membrane calcium pump isoform 1, plasma membrane calcium-ATPase, PMCA1kb, PMCA1plasma membrane calcium-transporting ATPase 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_001001323.1; NP_001673.2). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.03 - 0.1 ug/ml, Peptide ELISA Detection limit 1:64000 | |
| NP_001001323.1 | |
| ATP2B1 | |
| Peptide with sequence C-KQDGAIENRNKAKAQD corresponding to internal region according to NP_001673.2. | |
| 0.1 mg | |
| Cellular Markers | |
| 490 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction