Learn More
PLXDC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12285-1-AP-150UL
 View more versions of this product
Product Code. 16809913
  • 410.00 € / 150µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
May play a role in tumor angiogenesis.
Specifications
Specifications
PLXDC2
Polyclonal
Unconjugated
PLXDC2
plexin domain containing 2, PLXDC2, TEM7R
Rabbit
Antigen Affinity Chromatography
RUO
361282, 67448, 84898
-20°C
Liquid
Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6UX71, Q9DC11
Plxdc2
PLXDC2 Fusion Protein Ag2940
150 μL
Primary
Human, Mouse, Rat