missing translation for 'onlineSavingsMsg'
Learn More

PLX-4720, MedChemExpress

Product Code. 16424067
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16424067

Brand: Medchem Express HY51424

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PLX-4720 is a potent and selective inhibitor of B-RafV600E with IC50 of 13 nM in a cell-free assay, equally potent to c-Raf-1(Y340D and Y341D mutations), and 10-fold selectivity for B-RafV600E than wild-type B-Raf.

PLX-4720 is a potent and selective inhibitor of B-RafV600E with IC50 of 13 nM in a cell-free assay, equally potent to c-Raf-1(Y340D and Y341D mutations), and 10-fold selectivity for B-RafV600E than wild-type B-Raf.

Specifications

Chemical Name or Material PLX-4720
CAS 918505-84-7
Color White
Purity Grade Notes Research
Molecular Formula C17H14ClF2N3O3S
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (241.65 mM; Need ultrasonic)
SMILES O=S(NC1=CC=C(C(C(C2=CNC3=NC=C(C=C23)Cl)=O)=C1F)F)(CCC)=O
Molecular Weight (g/mol) 413.83
Formula Weight 413.83
Percent Purity 98.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.