missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLpro inhibitor, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
PLpro inhibitor is a potent inhibitor of papain-like protease (PLpro) with an IC50 of 2.6 μM. PLpro inhibitor inhibits SARS-CoV-2 PLpro with an IC50 of 5.0 μM and an EC50 of 21.0 μM.
Specifications
Specifications
| Chemical Name or Material | PLpro inhibitor |
| Physical Form | Solid |
| CAS | 1093070-14-4 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H22N2O2 |
| For Use With (Application) | COVID-19-anti-virus |
| Solubility Information | DMSO : ≥ 42 mg/mL (121.24 mM) |
| SMILES | O=C(C1=CC(NC(C)=O)=CC=C1C)N[C@H](C)C2=C(C=CC=C3)C3=CC=C2 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction