missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLEKHG2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15566762
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15566762 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15566762 Supplier Invitrogen™ Supplier No. PA545008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: DLTIPKHRHL LQAKNQEEKR LWIHCLQRLF FENHPASIPA KAKQVLLENS Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%; Zebrafish: 100%.

PLEKHG2 may be a transforming oncogene with exchange activity for CDC42. PLEKHG2 may be a guanine-nucleotide exchange factor (GEF) for RAC1 and CDC42.PLEKHG2 activated by the binding to subunits beta and gamma of the heterotrimeric guanine nucleotide-binding protein (G protein).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHG2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PLEKHG2
Gene Accession No. Q9H7P9
Gene Alias AI194308; ARHGEF42; Clg; common site lymphoma/leukemia GEF; Common site lymphoma/leukemia guanine nucleotide exchange factor; common-site lymphoma/leukemia GEF; common-site lymphoma/leukemia guanine nucleotide exchange factor; Cslg; CTB-60E11.4; DKFZp667J2325; FLJ00018; FLJ22458; FLJ38638; LDAMD; LOW QUALITY PROTEIN: pleckstrin homology domain-containing family G member 2; PH domain-containing family G member 2; pleckstrin homology and RhoGEF domain containing G2; pleckstrin homology domain containing, family G (with RhoGef domain) member 2; pleckstrin homology domain-containing family G member 2; Plekhg2; RGD1310112
Gene Symbols PLEKHG2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PLEKHG2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64857
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.