missing translation for 'onlineSavingsMsg'
Learn More

Pleconaril, MedChemExpress

Product Code. p-200095983
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497978 10 mg 10mg
16471296 200 mg 200mg
16463907 50 mg 50mg
16437766 100 mg 100mg
16430448 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497978 Supplier Medchem Express Supplier No. HY19952

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pleconaril is a capsid inhibitor used previously to treat enterovirus infections.

Pleconaril is a capsid inhibitor used previously to treat enterovirus infections.

Specifications

Chemical Name or Material Pleconaril
CAS 153168-05-9
Color White
Purity Grade Notes Research
Molecular Formula C18H18F3N3O3
Quantity 10 mg
Synonym VP 63843 Win 63843
Solubility Information DMSO : 100 mg/mL (262.23 mM; Need ultrasonic)
SMILES FC(C1=NC(C2=CC(C)=C(OCCCC3=CC(C)=NO3)C(C)=C2)=NO1)(F)F
Molecular Weight (g/mol) 381.35
Formula Weight 381.35
Percent Purity 99.35%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.