Learn More
Invitrogen™ PLCB2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5102006
Product Code. 16608484
  • 489.00 € / 100µL
Please to purchase this item. Need a web account? Register with us today!
Description
Description
Antibody detects endogenous levels of total PLCB2.
PLCB2 is the production of the second messenger molecules diacylglycerol (DAG) and inositol 1,4,5-trisphosphate (IP3) is mediated by activated phosphatidylinositol-specific phospholipase C enzymes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PLCB2
Polyclonal
Unconjugated
PLCB2
1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-2; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-2; AI550384; B230205M18Rik; B230399N12; Phosphoinositide phospholipase C-beta-2; phospholipase C beta 2; phospholipase C, beta 2; phospholipase C-beta-2; PKC beta II; PLC beta-2; PLCB2; plcb2 {ECO:0000250; PLCbeta2; PLC-beta2; PLC-beta-2; PLCbeta-2; UniProtKB:Q00722}
Rabbit
Affinity chromatography
RUO
18796, 5330, 85240
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
A3KGF7, O89040, Q00722
PLCB2
A synthesized peptide derived from human PLCB2(Accession Q00722), corresponding to amino acid residues S341-I391.
100 μL
Primary
Human, Mouse, Rat