Learn More
Platinum foil, 0.25mm (0.01in) thick, Premion™, 99.99% (metals basis)
Product Code. 11365879
Click to view available options
Quantity:
50 x 50 mm
Unit Size:
Each
Product Code. 11365879
Brand: Thermo Scientific Alfa Aesar 011508.FI
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
CAS: 7440-06-4
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 6-4-7440
Quantity 50 x 50 mm
Percent Purity 99.99%