missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibidi μ-Plate 96 Well Black ibiTreat
Description
- A black 96 well plate with flat and clear bottom for high throughput applications in cell-based assays
- bottom material: #1.5 Polymer Coverslip
- tissue culture treated
- sterilized
Specifications
Specifications
| Color | Black |
| Product Type | Well Plate |
| No. of Wells | 96 |
| Sterility | Sterile |
| Quantity | 15 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?