Learn More
Sperian™ Honeywell™ Plastic RD40 Filters
Product Code. 12663153
Change view
Click to view available options
For Use With (Application):
Dust and particulates protection
Gas vapor and particulates protection - A2B2E2K2 P3 large capacity for long duration work
Gas/vapor and particulates protection - AX large capacity for long duration work
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) unitSize
12663153 Gas/vapor and particulates protection - AX large capacity for long duration work Each
12653153 Gas vapor and particulates protection - A2B2E2K2 P3 large capacity for long duration work Each
12613143 Dust and particulates protection Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12663153 Supplier Sperian™ Supplier No. 1788161
Only null left
Add to basket
Add to basket
Protects against a multitude of risks encountered in many environments. Sperian™ Honeywell™ Plastic RD40 Filters are positioned low to prevent obstruction in the field of vision.
  • Body made of plastic provides optimum resistance
  • Low breathing resistance
  • Available in models to protect against airborne particulates, biohazards, chemicals, contamination and gas, vapors and smoke
TRUSTED_SUSTAINABILITY
Specifications
Cartridge Type Filter
Certifications/Compliance E.C. Declaration of Conformity- EC Category PPE- 3, Quality Assurance- ISO 9001 / 2000, EC Certificate Number- 0070-236-079-03-06-0006, EC Attestation- EC Attestation, EC Attestation Number- 0070-236-079-03-06-0006
Connection Type RD40
Material ABS material with plastic cover - High activated charcoal
Product Type Plastic Filters
For Use With (Application) Gas/vapor and particulates protection - AX large capacity for long duration work
For Use With (Equipment) Full-Face Mask Equipped With An RD40 Connector