Learn More
Sperian™ Honeywell™ Plastic RD40 Filters
Description
- Body made of plastic provides optimum resistance
- Low breathing resistance
- Available in models to protect against airborne particulates, biohazards, chemicals, contamination and gas, vapors and smoke
Specifications
Specifications
| Cartridge Type | Filter |
| Certifications/Compliance | E.C. Declaration of Conformity- EC Category PPE- 3, EC Certificate Number- 7793 A/13/8 PSA, EC Attestation- EC Attestation, EC Attestation Number- 7793 A/13/8 PSA, Certifications- EN 14387, Declaration of conformity DEKRA - 1788155 |
| Connection Type | RD40 |
| Material | ABS material with plastic cover - Activated and/or impregnated charcoal |
| Product Type | Plastic Filters |
| For Use With (Application) | Gas vapor and particulates protection - A2B2E2K2 P3 large capacity for long duration work |