Learn More
Sperian™ Honeywell™ Plastic RD40 Filters
Product Code. 12613143
Change view
Click to view available options
For Use With (Application):
Dust and particulates protection
Gas vapor and particulates protection - A2B2E2K2 P3 large capacity for long duration work
Gas/vapor and particulates protection - AX large capacity for long duration work
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) unitSize
12613143 Dust and particulates protection Each
12653153 Gas vapor and particulates protection - A2B2E2K2 P3 large capacity for long duration work Each
12663153 Gas/vapor and particulates protection - AX large capacity for long duration work Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12613143 Supplier Sperian™ Supplier No. 1786000
Only null left
Add to basket
Edge
Add to basket
Protects against a multitude of risks encountered in many environments. Sperian™ Honeywell™ Plastic RD40 Filters are positioned low to prevent obstruction in the field of vision.
  • Body made of plastic provides optimum resistance
  • Low breathing resistance
  • Available in models to protect against airborne particulates, biohazards, chemicals, contamination and gas, vapors and smoke
TRUSTED_SUSTAINABILITY
Specifications
Cartridge Type Filter
Certifications/Compliance E.C. Declaration of Conformity- EC Category PPE- 3, Quality Assurance- ISO 9001 / 2000, EC Certificate Number- 0070-003-079-06-98-0039, EC Attestation- EC Attestation, EC Attestation Number- 0070-003-079-06-98-0039, Certifications- EN 14387, EN 143, EN 12; EN 143 : 1998 type P3
Connection Type RD40
Material ABS Material With Plastic Cover
Product Type Plastic Filters
For Use With (Application) Dust and particulates protection
For Use With (Equipment) Full-Face Mask And Half-Mask Equipped With An Rd40 Connector