missing translation for 'onlineSavingsMsg'
Learn More

Plakophilin-3 Antibody (23E3/4), DyLight 488, Novus Biologicals™

Product Code. 18066876 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18066876 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18066876 Supplier Novus Biologicals Supplier No. NBP197674G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Plakophilin-3 Monoclonal specifically detects Plakophilin-3 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plakophilin-3
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 23E3/4
Conjugate DyLight 488
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Alias plakophilin 3, plakophilin-3
Gene Symbols PKP3
Host Species Mouse
Immunogen Clone 23E3/4 is a mouse monoclonal IgG2b antibody derived by fusion of mouse myeloma cells with spleen cells from a mouse immunized with a synthetic peptide corresponding to amino acid residues 570-589 of human plakophilin-3 coupled to keyhole limpet hemocyanin via an additional cysteine residue at the N-terminus (CFTPQSRRLRELPLAADALTF).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11187
Target Species Human, Mouse, Canine
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.