missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLAG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18018-1-AP-20UL
This item is not returnable.
View return policy
Description
Pleomorphic adenoma gene 1 encodes a zinc finger protein with 2 putative nuclear localization signals. PLAG1, which is developmentally regulated, has been shown to be consistently rearranged in pleomorphic adenomas of the salivary glands. PLAG1 is activated by the reciprocal chromosomal translocations involving 8q12 in a subset of salivary gland pleomorphic adenomas. Three transcript variants encoding two different isoforms have been found for this gene.Specifications
| PLAG1 | |
| Polyclonal | |
| Unconjugated | |
| PLAG1 | |
| PLAG1, pleiomorphic adenoma gene 1, PSA, SGPA, Zinc finger protein PLAG1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 297804, 5324, 56711 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunoprecipitation, Western Blot, Immunofluorescence | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5U2T6, Q6DJT9, Q9QYE0 | |
| PLAG1 | |
| PLAG1 Fusion Protein Ag12566 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction