missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PKC epsilon Polyclonal Antibody
GREENER_CHOICE

Product Code. 16679044
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16679044 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16679044 Supplier Invitrogen™ Supplier No. PA5102580

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PKC epsilon.

The PKC family of serine/threonine kinases, including PRKCE (PKC epsilon), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II, and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. Protein kinase C (PKC) can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKC epsilon
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PRKCE
Gene Accession No. P09216, P16054, Q02156
Gene Alias 5830406C15Rik; MGC125656; MGC125657; nPKC-epsilon; PKC e; PKC[e]; Pkce; Pkcea; PKCepsilon; PRKCE; PRKCH; protein kinase C; protein kinase C epsilon; protein kinase C epsilon type; protein kinase C, epsilon; protein kinase C, eta; R75156
Gene Symbols PRKCE
Host Species Rabbit
Immunogen A synthesized peptide derived from human PRKCE(Accession Q02156), corresponding to amino acid residues P683-E733.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18754, 29340, 5581
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.