Learn More
PITX2 Mouse anti-Human, Unconjugated, Clone: 1E3B1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67201-1-IG-20UL
Description
The PITX2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PITX2. This antibody recognizes human, mouse, rat antigen. The PITX2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PITX2 | |
| Monoclonal | |
| 800 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, ELISA | |
| 1E3B1 | |
| Unconjugated | |
| 20 μL | |
| 5308 | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.