Learn More
Pitofenone hydrochloride, MedChemExpress
Product Code. 16487097
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16487097
Brand: Medchem Express HY110389
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Pitofenone hydrochloride, a spasmolytic compound, inhibits the acetylcholinesterase (AChE) activity from bovine erythrocytes and from electric eel with Kis of 36 and 45 μM, respectively.
Pitofenone hydrochloride, a spasmolytic compound, inhibits the acetylcholinesterase (AChE) activity from bovine erythrocytes and from electric eel with Kis of 36 and 45 μM, respectively.
Specifications
Chemical Name or Material Pitofenone hydrochloride
CAS 1248-42-6
Color White
Purity Grade Notes Research
Molecular Formula C22H26ClNO4
Quantity 5 mg
Solubility Information H2O : 100 mg/mL (247.59 mM; Need ultrasonic) ∣DMSO : 83.33 mg/mL (206.31 mM; Need ultrasonic)
SMILES Cl[H].O=C(C1=CC=C(C=C1)OCCN2CCCCC2)C3=C(C=CC=C3)C(OC)=O
Molecular Weight (g/mol) 403.9
Formula Weight 403.9
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation