missing translation for 'onlineSavingsMsg'
Learn More

Pitavastatin-d5 Sodium Salt, TRC

Product Code. 30412847
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412847 1 mg 1mg
30425372 10 mg 10mg
30426260 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30412847 Supplier Toronto Research Chemicals Supplier No. TRCP5310071MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pitavastatin-d5 Sodium Salt
Purity Grade Notes HPLC
Molecular Formula C25H18D5FNNaO4
Quantity 1 mg
Synonym (3R,5S,6E)-7-[2-(Cyclopropyl-d5)-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-6-heptenoic Acid Sodium Salt, (+)-(3R,5S,6E)-7-[2-(Cyclopropyl-d5)-4-(4-fluorophenyl)-3-quinolyl] -3,5-dihydroxy-6-heptenoic Acid Sodium Salt, NK 104-d5 Sodium Salt
InChI Formula InChI=1S/C25H24FNO4.Na/c26-17-9-7-15(8-10-17)24-20-3-1-2-4-22(20)27-25(16-5-6-16)21(24)12-11-18(28)13-19(29)14-23(30)31;/h1-4,7-12,16,18-19,28-29H,5-6,13-14H2,(H,30,31);/q;+1/p-1/b12-11+;/t18-,19-;/m1./s1/i5D2,6D2,16D;
SMILES O[C@@H](C[C@@H](O)CC(O[Na])=O)/C=C/C1=C(C2=CC=C(F)C=C2)C3=C(C=CC=C3)N=C1C4([2H])C([2H])([2H])C4([2H])[2H]
Molecular Weight (g/mol) 448.47
Percent Purity >95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.