missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pirin Polyclonal Antibody

Description
Immunogen sequence: SFIYTISGDV YIGPDDAQQK IEPHHTAVLG EGDSVQVENK DPKRSHFVLI AGEPLREPVI QHGPFVMNTN EEISQAILDF RNAKNGFERA KTWKSK Antigen sequence identity: Mouse 95%; Rat 94%.
This gene encodes a member of the cupin superfamily. The encoded protein is an Fe(II)-containing nuclear protein expressed in all tissues of the body and concentrated within dot-like subnuclear structures. Interactions with nuclear factor I/CCAAT box transcription factor as well as B cell lymphoma 3-encoded oncoprotein suggest the encoded protein may act as a transcriptional cofactor and be involved in the regulation of DNA transcription and replication. Alternatively spliced transcript variants have been described.
Specifications
Specifications
| Antigen | Pirin |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PIR |
| Gene Accession No. | O00625 |
| Gene Alias | 2310042L19Rik; iron-binding nuclear protein; PIR; Pirin; pirin (iron-binding nuclear protein); pirin like; pirinin; Pirnl; Probable quercetin 2,3-dioxygenase PIR; probable quercetinase |
| Gene Symbols | PIR |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?