missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pirh2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550251
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total PIRH2 protein.
The protein encoded by this gene has ubiquitin-protein ligase activity. This protein binds with p53 and promotes the ubiquitin-mediated proteosomal degradation of p53. This gene is oncogenic because loss of p53 function contributes directly to malignant tumor development. Transcription of this gene is regulated by p53. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| Pirh2 | |
| Polyclonal | |
| Unconjugated | |
| RCHY1 | |
| Androgen receptor N-terminal-interacting protein; androgen-receptor N-terminal-interacting protein; ARNIP; Chimp; CH-rich interacting match with PLAG1; CH-rich-interacting match with PLAG1; DKFZp586C1620; E3 ubiquitin-protein ligase Pirh2; h PRO1996; hPirh2; p53-induced protein with a RING-H2 domain; p53-induced RING-H2 protein; PIRH2; PIRH2E; PIRH2F; PRO1996; RCHY1; ring finger and CHY zinc finger domain containing 1; ring finger and CHY zinc finger domain containing 1, E3 ubiquitin protein ligase; RING finger and CHY zinc finger domain-containing protein 1; RING finger protein 199; RING-type E3 ubiquitin transferase RCHY1; RNF199; ZCHY; Zfp363; Zinc finger protein 363; zinc finger, CHY-type; Znf363 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25898 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q96PM5 | |
| RCHY1 | |
| Full length fusion protein of Human RCHY1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction