Learn More
Greiner Bio-One Pipette Tips, 200-1000 μL, Universal, Blue
Product Code. 10194621
Click to view available options
Packaging:
Bag
Easyload Rack
Quantity:
250/Bag, 5000/Box
60/Rack, 2400/Box
Unit Size:
Pack of 1000
Pack of 2400
Product Code. 10194621
Brand: Greiner Bio-One 740265
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from high-grade polypropylene for improved performance, fit, and stability
  • Manufactured from high-grade polypropylene for optimum performance and fit
  • Polypropylene has a high breaking strength, is dimensionally stable, heat-resistant up to approx. 40°C, i.e. is autoclavable, and cold-resistant down to −190°C
  • Free of heavy metals
  • Manufactured under production conditions certified as conforming to the ISO 9001 standard.
  • Tested for conformance with the international standard DIN ISO 8655
  • Recommended when pipettes from different manufacturers are used at the same time
  • Ideally suited for Brand™; Eppendorf™; Finnpipette™; Gilson™; Socorex™
  • Volume: 200 μL - 1000 μL
  • Graduated
  • Matching rack Cat.-No. 941 325
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 200 to 1000 μL
Volume (Metric) Dispensing 200 to 1000 μL
Sterility Non-sterile
Color Blue
Autoclavable Autoclavable
Tip Style Universal
Packaging Easyload Rack
Quantity 60/Rack, 2400/Box
Graduated Yes
Material Polypropylene