Learn More
Hirschmann™ Pipette tips
Product Code. 10395031
Change view
Click to view available options
Volume (Metric):
0.2 to 10 μL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10395031 0.2 to 10 μL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10395031 Supplier Hirschmann™ Supplier No. 9490101
Only null left
Add to basket
Add to basket
Constructed from polypropylene. Hirschmann™ Pipette tips are for use with the labopette™
  • Loose in bag
  • Free of cadium
  • Certified conformity
  • Polypropylene
Specifications
Volume (Metric) 0.2 to 10 μL
Color Clear
Material Polypropylene