Learn More
Fisherbrand™ Single-Channel Pipette
Aspirates and dispenses viscous liquids accurately and reliably. Fisherbrand™ Single-Channel Pipette include a lock counter to prevent accidental volume changes. Lightweight, ergonomic design minimizes repetitive strain injuries.
Brand:  Fisherbrand™
52.21 EUR valid until 2023-10-01
Use promo code "20670" to get your promotional price.
Additional Details : Weight : 0.20700kg
 View more versions of this product
Product Code. 10063722
  • 142.00 € / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Accurate and reliable pipetting
  • Reverse pipetting option for accurate and precise aspirating and dispensing of viscous liquids
  • Innovative soft grip prevents transfer of heat to internal components, ensuring accurate pipette operation
  • Lock counter mechanism of variable volume pipettes prevents volume changes while pipetting
  • Comfortable and effortless pipetting - light weight / ergonomic design for easy operation prevents hand stress. Ergonomic low plunger force minimizes RSI (repetitive strain injury)
  • Pipette is fully autoclavable for sterile handling
  • Easy and quick maintenance and adjustment with the included disassembly and calibration tool
  • No loss of sample - blow out system allows last drop dispensing
  • Meets following standards: DIN EN ISO 8655-1 | DIN EN ISO 8655-2 | DIN EN ISO 8655-6 | EU Directive 98/79/EG
Specifications
Specifications
Autoclavable
Sterile
2 to 20 μL
0.50μL
2 Years
1
Manual Pipette
Lock counter mechanism, disassembly and calibration tool, sample bag with tips, grease
1 Each