Learn More
VITLAB™ Pipeo Pipette Controller
Brand:  VITLAB™ 1631500
Additional Details : Weight : 0.01000kg
Product Code. 10736033
  • 414.00 € / Each
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
Description
Description
Specifications
Specifications
Rechargeable
VITLAB pipeo&trade, charger (100 - 240 V, 50/60 Hz), 4 interchangeable plugs (EU, UK, US/J, AUS), rechargeable battery, one battery compartment cover, two replacement membrane filters (0.2 um)
Interchangeable (EU, UK, US/J, AUS)
100/240 V, 50/60 Hz