Learn More
PIP5K1A Mouse anti-Human, Unconjugated, Clone: 3E5G12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67483-1-IG-150UL
Description
The PIP5K1A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PIP5K1A. This antibody recognizes human antigen. The PIP5K1A antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| PIP5K1A | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, ELISA | |
| 3E5G12 | |
| Unconjugated | |
| 150 μL | |
| 8394 | |
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.