Learn More
Pinoxaden Despivoloyl (>90%), TRC
Specifications
Specifications
| Chemical Name or Material | Pinoxaden metabolite 1 NOA 407854 |
| CAS | 314020-44-5 |
| Molecular Formula | C18 H24 N2 O3 |
| Quantity | 10 mg |
| Synonym | 8-(2,6-Diethyl-4-methylphenyl)tetrahydro-7H-pyrazolo[1,2-d][1,4,5]oxadiazepine-7,9(8H)-dione, 8-(2,6-Diethyl-4-methylphenyl)tetrahydropyrazolo[1,2-d][1,4,5]oxadiazepine-7,9-dione, Pinoxaden metabolite M2 NOA 407854 |
| InChI Formula | InChI=1S/C18H24N2O3/c1-4-13-10-12(3)11-14(5-2)15(13)16-17(21)19-6-8-23-9-7-20(19)18(16)22/h10-11,16H,4-9H2,1-3H3 |
| SMILES | CCc1cc(C)cc(CC)c1C2C(=O)N3CCOCCN3C2=O |
| IUPAC Name | 8-(2,6-diethyl-4-methylphenyl)-1,2,4,5-tetrahydropyrazolo[1,2-d][1,4,5]oxadiazepine-7,9-dione |
| Molecular Weight (g/mol) | 316.3948 |
| Formula Weight | 316.1787 |