missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PIGY Polyclonal antibody specifically detects PIGY in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | PIGY |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 500 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Phosphatidylinositol-glycan biosynthesis class Y protein, PIG-Y |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human PIG-Y. The immunogen is located within the last 50 amino acids of PIG-Y. Amino Acid Squence: ARMTRQSKKQEEVEQR |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?