missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIDD Antibody (Lise-1), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11673PE
This item is not returnable.
View return policy
Description
PIDD Monoclonal antibody specifically detects PIDD in Mouse samples. It is validated for Western Blot
Specifications
| PIDD | |
| Monoclonal | |
| PE | |
| leucine-rich repeat and death domain-containing protein, leucine-rich repeats and death domain containing, MGC16925, p53-induced protein with a death domain, PIDD | |
| Mouse PIDD (Death Domain). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 55367 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot | |
| Lise-1 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction