Learn More
Pico145, MedChemExpress
Product Code. 16463398
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463398 10 mg 10mg
16459926 50 mg 50mg
16444636 100 mg 100mg
16426127 5 mg 5mg
16422756 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16463398 Supplier Medchem Express Supplier No. HY101507
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Pico145 (HC-608) is a remarkable inhibitor of TRPC1/4/5 channels, inhibits (−)-englerin A-activated TRPC4/TRPC5 channels, with IC50s of 0.349 and 1.3 nM in cells, and shows no effect on TRPC3, TRPC6, TRPV1, TRPV4, TRPA1, TRPM2, TRPM8.
Pico145 (HC-608) is a remarkable inhibitor of TRPC1/4/5 channels, inhibits (−)-englerin A-activated TRPC4/TRPC5 channels, with IC50s of 0.349 and 1.3 nM in cells, and shows no effect on TRPC3, TRPC6, TRPV1, TRPV4, TRPA1, TRPM2, TRPM8.
Specifications
Chemical Name or Material Pico145
CAS 1628287-16-0
Color Pink
Purity Grade Notes Research
Molecular Formula C23H20ClF3N4O5
Quantity 10 mg
Synonym HC-608
Solubility Information DMSO : ≥ 100 mg/mL (190.52 mM)
SMILES O=C1C2=C(N=C(OC3=CC=CC(OC(F)(F)F)=C3)N2CC4=CC=C(Cl)C=C4)N(C)C(N1CCCO)=O
Molecular Weight (g/mol) 524.88
Formula Weight 524.88
Percent Purity 98.62%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death