Description
PICK1 Polyclonal antibody specifically detects PICK1 in Human samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | PICK1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Formulation | PBS |
| Gene Alias | alpha binding protein, dJ1039K5, MGC15204, PICK, PRKCA-binding protein, PRKCABPprotein interacting with PRKCA, Protein interacting with C kinase 1, protein interacting with PRKCA 1, Protein kinase C-alpha-binding protein |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 300-400 of human PICK1 (NP_036539.1).,, Sequence:, YLSYCLKVKEMDDEEYSCIALGEPLYRVSTGNYEYRLILRCRQEARARFSQMRKDVLEKMELLDQKHVQDIVFQLQRLVSTMSKYYNDCYAVLRDADVFPI |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |