Learn More
Piceatannol, MedChemExpress
Product Code. 16405728
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
Product Code. 16405728
Brand: Medchem Express HY13518
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Piceatannol is a well-known Syk inhibitor and reduces the expression of iNOS induced by TNF.
Piceatannol is an effective agent for research of acute lung injury (ALI). Piceatannol is a naturally occurring polyphenolic stilbene found in various fruits and vegetables and exhibits anticancer and anti-inflammatory properties. Piceatannol induces apoptosis in DLBCL cell lines. Piceatannol induces autophagy and apoptosis in MOLT-4 human leukemia cells.
Specifications
Chemical Name or Material Piceatannol
CAS 10083-24-6
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C14H12O4
Quantity 10 mg
Synonym Astringenin trans-Piceatannol
Solubility Information DMSO : ≥ 31 mg/mL (126.92 mM) ∣H2O : 1 mg/mL (4.09 mM; Need ultrasonic)
SMILES OC1=CC=C(/C=C/C2=CC(O)=CC(O)=C2)C=C1O
Molecular Weight (g/mol) 244.24
Formula Weight 244.24
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease