missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI 3-Kinase p85 beta Antibody (T15), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65590B
This item is not returnable.
View return policy
Description
PI 3-Kinase p85 beta Monoclonal antibody specifically detects PI 3-Kinase p85 beta in Human, Mouse, Rat, Bovine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
| PI 3-Kinase p85 beta | |
| Monoclonal | |
| Biotin | |
| P85B, phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta, phosphatidylinositol 3-kinase regulatory subunit beta, phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 2 (p85 beta), phosphoinositide-3-kinase, regulatory subunit 2 (beta), phosphoinositide-3-kinase, regulatory subunit 2 (p85 beta), phosphoinositide-3-kinase, regulatory subunit, polypeptide 2 (p85 beta), PI3K regulatory subunit beta, PI3-kinase regulatory subunit beta, PI3-kinase subunit p85-beta, ptdIns-3-kinase regulatory subunit beta, ptdIns-3-kinase regulatory subunit p85-beta | |
| Balculovirus expressed (Recombinant) Bovine P85 beta subunit of P13 kinase. | |
| 0.1 mL | |
| Autophagy, Signal Transduction | |
| 5296 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| T15 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction