missing translation for 'onlineSavingsMsg'
Learn More

PI 3-Kinase C2 beta Antibody (OTI3B1), DyLight 755, Novus Biologicals™

Product Code. 18139606 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18139606 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18139606 Supplier Novus Biologicals Supplier No. NBP273395IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PI 3-Kinase C2 beta Monoclonal specifically detects PI 3-Kinase C2 beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI 3-Kinase C2 beta
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3B1
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C2-PI3KDKFZp686G16234, EC 2.7.1, EC 2.7.1.154, phosphatidylinositol 3-kinase C2 domain-containing beta polypeptide, phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit beta, Phosphoinositide 3-kinase-C2-beta, phosphoinositide-3-kinase, class 2, beta polypeptide, PI3K-C2beta, PI3K-C2-beta, PTDINS-3-kinase C2 beta, PtdIns-3-kinase C2 subunit beta
Gene Symbols PIK3C2B
Host Species Mouse
Immunogen Full length recombinant protein of human PI 3-Kinase C2 beta (NP_002637) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5287.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.