missing translation for 'onlineSavingsMsg'
Learn More

Phyllochromenol, TRC

Product Code. 30005743
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005743 10 mg 10mg
30137348 2.5 mg 2.5mg
30133584 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005743 Supplier Toronto Research Chemicals Supplier No. TRCP41303510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19274-66-9
Formula Weight 450.3498
InChI Formula InChI=1S/C31H46O2/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-20-31(6)21-19-26-25(5)29(32)27-17-7-8-18-28(27)30(26)33-31/h7-8,17-19,21-24,32H,9-16,20H2,1-6H3
IUPAC Name 2,5-dimethyl-2-(4,8,12-trimethyltridecyl)benzo[h]chromen-6-ol
Molecular Formula C31H46O2
Molecular Weight (g/mol) 450.7
SMILES CC(C)CCCC(C)CCCC(C)CCCC1(C)Oc2c(C=C1)c(C)c(O)c3ccccc23
Synonym 2,5-Dimethyl-2-(4,8,12-trimethyltridecyl)-2H-benzo[h]chromen-6-ol
Chemical Name or Material Phyllochromenol

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.