missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHOX2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540121
This item is not returnable.
View return policy
Description
Recommended positive controls: IMR32, SK-N-AS, and SH-SY5Y cells. Predicted reactivity: Mouse (100%), Xenopus laevis (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The DNA-associated protein encoded by this gene is a member of the paired family of homeobox proteins localized to the nucleus. The protein functions as a transcription factor involved in the development of several major noradrenergic neuron populations and the determination of neurotransmitter phenotype. The gene product is linked to enhancement of second messenger-mediated activation of the dopamine beta-hydroylase, c-fos promoters and several enhancers, including cyclic amp-response element and serum-response element.
Specifications
| PHOX2B | |
| Polyclonal | |
| Unconjugated | |
| PHOX2B | |
| dilated pupils 1; Dilp1; GENA 269; LOW QUALITY PROTEIN: paired mesoderm homeobox protein 2B; NBLST2; NBPhox; neuroblastoma paired-type homeobox protein; neuroblastoma Phox; paired like homeobox 2B; paired mesoderm homeobox 2b; paired mesoderm homeobox protein 2B; Paired-like homeobox 2B; PHOX2B; PHOX2B homeodomain protein; Pmx2b | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8929 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1.42 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q99453 | |
| PHOX2B | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human PHOX2B. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction