missing translation for 'onlineSavingsMsg'
Learn More

WTW™ photoLab™ S6 Filter Photometer

The photoLab™ S6 filter photometer with 6 wavelengths combines lab precision with the highest comfort and speeds for water analytics.

Brand:  WTW™ 250013D

Additional Details : Weight : 3.50000kg

Product Code. 10180891

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • 6 wavelengths for quick tests with 16mm round cuvettes
  • Reference beam optics
  • Barcode detection
  • AQA/IQC multi-level
  • Uncomplicated in its operation with occasional measurements
  • Series examinations with high test throughput at highest speeds
  • 500 data sets
  • Applications include water analytics, galvanization and in the food industry
Specifications

Specifications

LCD
10.62 in.
Water Analytics, Galvanization, Food Industry
230 V
140 mm
340, 445, 525, 550, 605, 690nm
10.23 in.
±2nm
270 mm
5.51 in.
50 Hz
Reference beam with filter
260 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Promotions

Promotions