missing translation for 'onlineSavingsMsg'
Learn More

Phosphoserine Monoclonal Antibody (PSR-45), Invitrogen™

Product Code. 11530982
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11530982 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11530982 Supplier Invitrogen Supplier No. MA190649

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-90649 detects Phosphoserine from various samples. MA1-90649 has been successfully used in ELISA, Western blot applications. The MA1-90649 immunogen is: Recombit fragment: (pS) conjugated to KLH. For indirect ELISA use a dilution of 1:4000.

Phosphoserine is the phosphoric acid ester of serine. As a constituent (residue) of proteins, its side chain can undergo O-linked glycosylation. This might be important in explaining some of the devastating consequences of diabetes. It is one of three amino acid residues that are commonly phosphorylated by kinases during cell signalling in eukaryotes. Phosphorylated serine residues are often referred to as phosphoserine. Serine proteases are a common type of protease. Serine, an organic compound, is one of the 20 amino acids commonly found in animal proteins. Only the L-stereoisomer appears in mammalian protein. It is not essential to the human diet, since it can be synthesized in the body from other metabolites, including glycine. [National Center for Biotechnology Information. PubChem Compound Database; CID=68841]
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphoserine
Applications ELISA, Western Blot
Classification Monoclonal
Clone PSR-45
Conjugate Unconjugated
Gene Alias pSer; Serine; Phospho-Serine; O-phospho-L-serine; Dexfosfoserine; Fosforina; L-O-Phosphoserine; O-Phosphoserine; C3H8NO6P
Host Species Mouse
Immunogen Recombit fragment: (pS) conjugated to KLH.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.