Learn More
Phosphoric Acid Tripropyl Ester-d21, TRC
Product Code. 30412806
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412806 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30412806 Supplier Toronto Research Chemicals Supplier No. TRCP359397100MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Phosphoric Acid Tripropyl Ester-d21
CAS 1219794-92-9
Molecular Formula C9 D21 O4 P
Quantity 100 mg
Synonym 1-Propan-1,1,2,2,3,3,3-d7-ol','1,1',1''-phosphate, Tri-n-propyl Phosphate D21, Tripropyl Phosphate-d21, 1-Propan-1,1,2,2,3,3,3-d7-ol','1,1',1''-phosphate, Tri-n-propyl Phosphate D21
InChI Formula InChI=1S/C9H21O4P/c1-4-7-11-14(10,12-8-5-2)13-9-6-3/h4-9H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2,7D2,8D2,9D2
SMILES [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])OP(=O)(OC([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])OC([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
IUPAC Name tris(1,1,2,2,3,3,3-heptadeuteriopropyl) phosphate
Molecular Weight (g/mol) 245.36
Formula Weight 245.2496